Bis[2-(diisopropylphosphino)ethyl]amine 10% in THF

Supplier: TCI America
Danger

B5712-1G B5712-5G
TCB5712-1GEA 132 USD
TCB5712-1G TCB5712-5G
Bis[2-(diisopropylphosphino)ethyl]amine 10% in THF
Bis[2-(diisopropylphosphino)ethyl]amine
Formula: C₁₆H₃₇NP₂
Flash Pt: -17 °C
MDL Number: MFCD17018778
CAS Number: 131890-26-1
UN: 2056
ADR: 3,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (Neutralization titration) 9.5 to 11.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR