2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA) ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
Vadimezan, ASA-404, DMXAA, 2-(5,6-Dimethyl-9-oxo-9H-xanthen-4-yl)acetic acid, 2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid
D5235-250MG
D5235-50MG
TCD5235-250MGEA
455
USD
TCD5235-250MG
TCD5235-50MG
2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA) ≥98.0% (by HPLC)
2-(5,6-Dimethyl-9-oxoxanthen-4-yl)acetic acid (DMXAA)
Formula:
C₁₇H₁₄O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD00870555 CAS Number: 117570-53-3 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...