1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane, 1,2-Bis[(2R,5R)-2,5-dimethylphospholano]ethane, 1,2-Bis[(2R,5R)-2,5-dimethylphospholan-1-yl]ethane, (R,R)-Me-BPE

B5985-100MG B5985-500MG
TCB5985-100MGEA 245 USD
TCB5985-100MG TCB5985-500MG
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane ≥95.0% (by GC)
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]ethane
Formula: C₁₄H₂₈P₂
MW: 258.326 g/mol
Boiling Pt: 67 °C
MDL Number: MFCD01073770
CAS Number: 129648-07-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR