1-Cyclopentylethanone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3585-1G C3585-5G
TCC3585-1GEA 57 USD
TCC3585-1G TCC3585-5G
1-Cyclopentylethanone ≥98.0% (by GC)
1-Cyclopentylethanone
Formula: C₇H₁₂O
Boiling Pt: 156 °C
MDL Number: MFCD00060799
CAS Number: 6004-60-0
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR