2-Bromo-6-hydrazinylpyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5120-1G B5120-5G
TCB5120-1GEA 57 USD
TCB5120-1G TCB5120-5G
2-Bromo-6-hydrazinylpyridine ≥98.0% (by GC, titration analysis)
2-Bromo-6-hydrazinylpyridine
Formula: C₅H₆BrN₃
MDL Number: MFCD09743754
CAS Number: 26944-71-8

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Potassium iodate Method) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR