1-(Boc-amino)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B6086-1G B6086-5G
TCB6086-1GEA 27.52 USD
TCB6086-1G TCB6086-5G
1-(Boc-amino)cyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Boc-amino)cyclobutanecarboxylic acid
Formula: C₁₀H₁₇NO₄
MDL Number: MFCD02682623
CAS Number: 120728-10-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 131.0 to 135.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR