5-Bromo-2-chloronicotinonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

B4870-1G B4870-5G
TCB4870-1GEA 55 USD
TCB4870-1G TCB4870-5G
5-Bromo-2-chloronicotinonitrile ≥98.0% (by GC)
5-Bromo-2-chloronicotinonitrile
Formula: C₆H₂BrClN₂
MDL Number: MFCD09801046
CAS Number: 405224-23-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR