1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene, 1,2-Bis((2R,5R)-2,5-dimethylphospholan-1-yl)benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholan-1-yl]benzene, 1,2-Bis[(2R,5R)-2,5-dimethylphospholano]benzene, (R,R)-Me-DUPHOS

B2436-100MG B2436-500MG
TCB2436-100MGEA 98 USD
TCB2436-100MG TCB2436-500MG
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene ≥98.0% (by GC)
1,2-Bis[(2R,5R)-2,5-dimethyl-1-phospholanyl]benzene
Formula: C₁₈H₂₈P₂
MW: 306.37 g/mol
MDL Number: MFCD00142336
CAS Number: 147253-67-6
Merck Index: 14,03466

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Specific rotation [a]20/D -470 to -520 deg (C=1, hexane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR