(4-Bromophenylethynyl)trimethylsilane

Supplier: TCI America
Warning

Synonyms: ((4-Bromophenyl)ethynyl)trimethylsilane

B6028-1G B6028-5G
TCB6028-1GEA 36 USD
TCB6028-1G TCB6028-5G
(4-Bromophenylethynyl)trimethylsilane
(4-Bromophenylethynyl)trimethylsilane
Formula: C₁₁H₁₃BrSi
Melting Pt: 62 °C
MDL Number: MFCD01863556
CAS Number: 16116-78-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR