cis-1,4-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B5146-1G B5146-5G
TCB5146-1GEA 368 USD
TCB5146-1G TCB5146-5G
cis-1,4-Bis(aminomethyl)cyclohexane ≥98.0% (by GC, titration analysis)
cis-1,4-Bis(aminomethyl)cyclohexane
Formula: C₈H₁₈N₂
Boiling Pt: 76 °C
Melting Pt: -9 °C
CAS Number: 10029-09-1
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR