4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Pentylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Amylbenzoic acid 4-Cyano-3-fluorophenyl ester, 4-Cyano-3-fluorophenyl 4-Pentylbenzoate, 4-Cyano-3-fluorophenyl 4-Amylbenzoate

C3555-1G C3555-5G
TCC3555-1GEA 30 USD
TCC3555-1G TCC3555-5G
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)
4-Cyano-3-fluorophenyl-4-n-pentylbenzoate
Formula: C₁₉H₁₈FNO₂
MW: 311.356 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11053363
CAS Number: 86786-89-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR