4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

C3512-1G C3512-5G
TCC3512-1GEA 60 USD
TCC3512-1G TCC3512-5G
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate ≥98.0% (by GC)
4-Cyano-3,5-difluorophenyl-4-n-pentylbenzoate
Formula: C₁₉H₁₇F₂NO₂
MDL Number: MFCD11053375
CAS Number: 123843-69-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR