4,5-Dibromo-1H-1,2,3-triazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

D5205-1G D5205-5G
TCD5205-1GEA 81 USD
TCD5205-1G TCD5205-5G
4,5-Dibromo-1H-1,2,3-triazole ≥98.0% (by titrimetric analysis)
4,5-Dibromo-1H-1,2,3-triazole
Formula: C₂HBr₂N₃
MDL Number: MFCD08062401
CAS Number: 15294-81-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR