3-Fluoro-4-cyanophenyl-4-ethylbenzoate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Cyano-3-fluorophenyl 4-Ethylbenzoate, 4-Ethylbenzoic acid 4-Cyano-3-fluorophenyl ester
C3540-25G
C3540-5G
TCC3540-25GEA
255
USD
TCC3540-25G
TCC3540-5G
3-Fluoro-4-cyanophenyl-4-ethylbenzoate ≥98.0% (by GC)
3-Fluoro-4-cyanophenyl-4-ethylbenzoate
Formula:
C₁₆H₁₂FNO₂ MW: 269.275 g/mol |
MDL Number:
MFCD00689047 CAS Number: 86776-50-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 77.0 to 81.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...