2-Bromoresorcinol ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B5737-1G B5737-5G
TCB5737-1GEA 39 USD
TCB5737-1G TCB5737-5G
2-Bromoresorcinol ≥98.0% (by titrimetric analysis)
2-Bromoresorcinol
Formula: C₆H₅BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD07779241
CAS Number: 6751-75-3

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystaline
Purity (GC) min. 97.0 %
Purity (Bromination Method) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR