(2-Bromoethoxy)-tert-butyldimethylsilane ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (2-Bromoethoxy)(tert-butyl)dimethylsilane

B4505-5G B4505-25G
TCB4505-5GEA 48 USD
TCB4505-5G TCB4505-25G
(2-Bromoethoxy)-tert-butyldimethylsilane ≥96.0% (by GC)
(2-Bromoethoxy)-tert-butyldimethylsilane
Formula: C₈H₁₉BrOSi
MW: 239.228 g/mol
Boiling Pt: 120 °C
Flash Pt: 73 °C
MDL Number: MFCD00209550
CAS Number: 86864-60-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR