1-Bromo-5-fluoropentane ≥98.0% (by GC)

Supplier: TCI America
Warning

B5998-25G B5998-5G
TCB5998-25GEA 142 USD
TCB5998-25G TCB5998-5G
1-Bromo-5-fluoropentane ≥98.0% (by GC)
1-Bromo-5-fluoropentane
Formula: C₅H₁₀BrF
Boiling Pt: 162 °C
MDL Number: MFCD01709395
CAS Number: 407-97-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR