Boc-Val-OMe ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Boc-L-valine methyl ester, Boc-Val-OMe, N-(tert-Butoxycarbonyl)-L-valine methyll ester, (S)-Methyl 2-[(tert-Butoxycarbonyl)amino]-3-methylbutanoate, (S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-methylbutanoate, N-Boc-L-valine methyll ester

B6020-25G B6020-5G
TCB6020-25GEA 229 USD
TCB6020-25G TCB6020-5G
Boc-Val-OMe ≥95.0% (by HPLC, titration analysis)
Boc-Val-OMe
Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
MDL Number: MFCD00672510
CAS Number: 58561-04-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Specific rotation [a]20/D +11.0 to +15.0 deg (C=1, CHCL3)
Acid Value max. 1.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR