H-Glu-otbu ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-4-Amino-5-(tert-butoxy)-5-oxopentanoic acid, L-Glutamic acid 1-tert-butyl ester, 1-tert-Butyl L-Glutamate, H-Glu-OtBu, L-Glutamic acid 1-tert-Butyl ester

B3000-25G B3000-5G
TCB3000-25GEA 426 USD
TCB3000-25G TCB3000-5G
H-Glu-otbu ≥98.0% (by titrimetric analysis)
H-Glu-otbu
Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Number: MFCD00038562
CAS Number: 45120-30-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 138.0 to 142.0 deg-C
Specific rotation [a]20/D +12.0 to +16.0 deg (C=1, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR