(S)-Methyl-2-acetamidopropanoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ac-Ala-OMe, N-Acetyl-L-alanine methyl ester

A3262-25G A3262-5G
TCA3262-25GEA 105 USD
TCA3262-25G TCA3262-5G
(S)-Methyl-2-acetamidopropanoate ≥98.0% (by GC)
(S)-Methyl-2-acetamidopropanoate
Formula: C₆H₁₁NO₃
MW: 145.158 g/mol
Boiling Pt: 90 °C
MDL Number: MFCD00038239
CAS Number: 3619-02-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -98.0 to -88.0 deg (C=1,H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR