Fmoc-Lys(Z)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-carbobenzoxy-L-lysine, Fmoc-Lys(Cbz)-OH, Fmoc-Lys(Z)-OH, Nalpha-Fmoc-Nepsilon-benzyloxycarbonyl-L-lysine, Nalpha-Fmoc-Nepsilon-Cbz-L-lysine

C3147-25G C3147-5G
TCC3147-25GEA 137 USD
TCC3147-25G TCC3147-5G
Fmoc-Lys(Z)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-Lys(Z)-OH
Formula: C₂₉H₃₀N₂O₆
MW: 502.567 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00065662
CAS Number: 86060-82-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C
Specific rotation [a]20/D -9.0 to -11.0 deg (C=2,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR