Cobalt naphthenate (Co ∼8%)

Supplier: TCI America
Danger

Synonyms: Naphthenic Acid Cobalt Salt

C3480-25G C3480-500G
TCC3480-25GEA 25 USD
TCC3480-25G TCC3480-500G
Cobalt naphthenate (Co ∼8%)
Cobalt naphthenate
Formula: C₂₂H₁₄CoO₄
MW: 401.28 g/mol
Flash Pt: 48 °C
MDL Number: MFCD02094763
CAS Number: 61789-51-3
EINECS: 263-064-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Gray to Dark purple to Black clear liquid to cloudy liquid
Cobalt 7.5 to 8.4 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR