4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Supplier: TCI America

A3327-1G A3327-200MG
TCA3327-1GEA 135 USD
TCA3327-1G TCA3327-200MG
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride
Formula: C₅H₁₂ClNO₂S
Melting Pt: 289 °C
MDL Number: MFCD11114559
CAS Number: 116529-31-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR