3-Chlorophenyl acetylene ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1-Chloro-3-ethynylbenzene, 3-Chlorophenylacetylene, (3-Chlorophenyl)acetylene

C3099-1G C3099-5G
TCC3099-1GEA 27 USD
TCC3099-1G TCC3099-5G
3-Chlorophenyl acetylene ≥97.0% (by GC)
3-Chlorophenyl acetylene
Formula: C₈H₅Cl
MW: 136.578 g/mol
Boiling Pt: 72 °C
Flash Pt: 62 °C
MDL Number: MFCD03839984
CAS Number: 766-83-6

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR