4-Bromophthalonitrile ≥97.0% (by GC)

Supplier: TCI America
Danger

B5509-1G B5509-5G
TCB5509-1GEA 86 USD
TCB5509-1G TCB5509-5G
4-Bromophthalonitrile ≥97.0% (by GC)
4-Bromophthalonitrile
Formula: C₈H₃BrN₂
MDL Number: MFCD00221794
CAS Number: 70484-01-4
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 138.0 to 142.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR