2-((tert-Butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
N-Boc-DL-alanine, N-(tert-Butoxycarbonyl)-DL-alanine, Boc-DL-Ala-OH
B4282-25G
B4282-5G
TCB4282-25GEA
55
USD
TCB4282-25G
TCB4282-5G
2-((tert-Butoxycarbonyl)amino)propanoic acid ≥98.0% (by HPLC, titration analysis)
2-((tert-Butoxycarbonyl)amino)propanoic acid
Formula:
C₈H₁₅NO₄ MW: 189.2 g/mol |
MDL Number:
MFCD00063124 CAS Number: 3744-87-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 110.0 to 114.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...