3-(Benzyloxy)cyclobutanone ≥95.0% (by GC)

Supplier: TCI America

B6029-1G B6029-200MG
TCB6029-1GEA 108 USD
TCB6029-1G TCB6029-200MG
3-(Benzyloxy)cyclobutanone ≥95.0% (by GC)
3-(Benzyloxy)cyclobutanone
Formula: C11H12O2
Boiling Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09755991
CAS Number: 30830-27-4

Order Now

Specification Test Results

Appearance Colorless to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR