tert-Butyl methyl(2-oxoethyl)carbamate ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: tert-Butyl methyll(2-oxoethyl)carbamate, Methyl(2-oxoethyl)carbamic acid tert-butyl ester, N-Boc-(methylamino)acetaldehyde, N-tert-Butoxycarbonyl-(methylamino)acetaldehyde

B5825-1G B5825-5G
TCB5825-1GEA 43 USD
TCB5825-1G TCB5825-5G
tert-Butyl methyl(2-oxoethyl)carbamate ≥96.0% (by GC)
tert-Butyl methyl(2-oxoethyl)carbamate
Formula: C₈H₁₅NO₃
MW: 173.21 g/mol
Boiling Pt: 89 °C
MDL Number: MFCD08064267
CAS Number: 123387-72-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %
Acid Value max. 5.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR