1-N-Cbz-4-aminopiperidine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-Amino-1-piperidinecarboxylic acid Benzyl ester, 4-Amino-1-benzyloxycarbonylpiperidine, 4-Amino-1-carbobenzoxypiperidine, Benzyl 4-aminopiperidine-1-carboxylate, 4-Amino-1-Cbz-piperidine, 1-N-Cbz-4-Aminopiperidine, Benzyl 4-Amino-1-piperidinecarboxylate

A2756-1G A2756-5G
TCA2756-1GEA 33 USD
TCA2756-1G TCA2756-5G
1-N-Cbz-4-aminopiperidine ≥98.0% (by GC)
1-N-Cbz-4-aminopiperidine
Formula: C₁₃H₁₈N₂O₂
MW: 234.30 g/mol
MDL Number: MFCD05863884
CAS Number: 120278-07-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR