2-(Bromomethyl)-5-(trifluoromethyl)furan ≥97.0% (by GC)
Supplier: TCI America
|
Danger
|
B5800-1G
B5800-200MG
TCB5800-1GEA
157
USD
TCB5800-1G
TCB5800-200MG
2-(Bromomethyl)-5-(trifluoromethyl)furan ≥97.0% (by GC)
2-(Bromomethyl)-5-(trifluoromethyl)furan
Formula:
C₆H₄BrF₃O MW: 228.9946 g/mol |
MDL Number:
MFCD03086219 CAS Number: 17515-77-4 UN: 2920 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...