1-Bromo-3-chloro-5-iodobenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

B4981-1G B4981-5G
TCB4981-1GEA 61 USD
TCB4981-1G TCB4981-5G
1-Bromo-3-chloro-5-iodobenzene ≥97.0% (by GC)
1-Bromo-3-chloro-5-iodobenzene
Formula: C₆H₃BrClI
MDL Number: MFCD07778998
CAS Number: 13101-40-1

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 84.0 to 88.4 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR