3-Bromo-5-methylaniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
B6015-25G
B6015-5G
TCB6015-25GEA
627
USD
TCB6015-25G
TCB6015-5G
3-Bromo-5-methylaniline ≥98.0% (by GC, titration analysis)
3-Bromo-5-methylaniline
Formula:
C7H8BrN MW: 186.05 g/mol Boiling Pt: 260 °C Melting Pt: 37 °C |
MDL Number:
MFCD11845553 CAS Number: 74586-53-1 EINECS: 277-932-1 |
Specification Test Results
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...