2-Bromo-4,5-difluorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B5722-25G B5722-5G
TCB5722-25GEA 233 USD
TCB5722-25G TCB5722-5G
2-Bromo-4,5-difluorobenzoic acid ≥98.0% (by GC, titration analysis)
2-Bromo-4,5-difluorobenzoic acid
Formula: C₇H₃BrF₂O₂
MDL Number: MFCD04973422
CAS Number: 64695-84-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR