2-(Benzyloxy)-1-ethanamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

B5880-1G B5880-5G
TCB5880-1GEA 30 USD
TCB5880-1G TCB5880-5G
2-(Benzyloxy)-1-ethanamine ≥98.0% (by GC, titration analysis)
2-(Benzyloxy)-1-ethanamine
Formula: C₉H₁₃NO
Boiling Pt: 100 °C
MDL Number: MFCD00235180
CAS Number: 38336-04-8
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR