tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate ≥97.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 1-(tert-Butoxycarbonyl)-4-(4-aminophenyl)piperidine, 1-Boc-4-(4-aminophenyl)piperidine, tert-Butyl 4-(4-aminophenyl)piperidine-1-carboxylate, 4-(4-Aminophenyl)piperidine-1-carboxylic acid tert-Butyl ester

B5832-1G B5832-200MG
TCB5832-1GEA 66 USD
TCB5832-1G TCB5832-200MG
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate ≥97.0% (by HPLC)
tert-Butyl-4-(4-aminophenyl)piperidine-1-carboxylate
Formula: C₁₆H₂₄N₂O₂
MW: 276.3740 g/mol
MDL Number: MFCD05861405
CAS Number: 170011-57-1

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR