2-Chloro-5-(chloromethyl)thiazole ≥98.0% (by GC)

Supplier: TCI America
Danger

C3295-5G C3295-25G
TCC3295-5GEA 25 USD
TCC3295-5G TCC3295-25G
2-Chloro-5-(chloromethyl)thiazole ≥98.0% (by GC)
2-Chloro-5-(chloromethyl)thiazole
Formula: C₄H₃Cl₂NS
MW: 168.0443 g/mol
Melting Pt: 35 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01073549
CAS Number: 105827-91-6
EINECS: 429-830-5
UN: 2922
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR