2,6-Dichloro-3-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
D5173-1G
D5173-5G
TCD5173-1GEA
93
USD
TCD5173-1G
TCD5173-5G
2,6-Dichloro-3-nitrobenzoic acid ≥98.0% (by titrimetric analysis)
2,6-Dichloro-3-nitrobenzoic acid
Formula:
C₇H₃Cl₂NO₄ |
MDL Number:
MFCD00075422 CAS Number: 55775-97-8 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 149.0 to 153.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...