5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

A3230-25G A3230-5G
TCA3230-25GEA 69 USD
TCA3230-25G TCA3230-5G
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥98.0% (by GC)
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile
Formula: C₁₁H₅Cl₂F₃N₄
MDL Number: MFCD00227537
CAS Number: 120068-79-3
EINECS: 421-240-6
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 149.0 to 153.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR