Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4,4'-Dimethoxy-5,6,5',6'-di(methylenedioxy)biphenyl-2,2'-dicarboxylic acid dimethyl ester, Dimethyl 4,4'-Dimethoxy-5,6,5',6'-di(methylenedioxy)biphenyl-2,2'-dicarboxylate, Bifendate, Dimethyl 7,7'-dimethoxy-[4,4'-bibenzo[d][1,3]dioxole]-5,5'-dicarboxylate
B6112-25G
B6112-5G
TCB6112-25GEA
115
USD
TCB6112-25G
TCB6112-5G
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate ≥98.0% (by GC)
Dimethyl-7,7'-dimethoxy[4,4'-bibenzo[d][1,3]dioxole]5,5'-dicarboxylate
Formula:
C₂₀H₁₈O₁₀ MW: 418.3509 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD01751431 CAS Number: 73536-69-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 179.0 to 183.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...