1-Benzyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Benzyl-1H-pyrazole-4-carboxaldehyde

B5404-1G B5404-200MG
TCB5404-1GEA 163 USD
TCB5404-1G TCB5404-200MG
1-Benzyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
1-Benzyl-1H-pyrazole-4-carbaldehyde
Formula: C₁₁H₁₀N₂O
Boiling Pt: 144 °C
MDL Number: MFCD02179567
CAS Number: 63874-95-3

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR