6-Bromochroman-4-one ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 6-Bromo-4-chromanone

B5843-1G B5843-5G
TCB5843-1GEA 19 USD
TCB5843-1G TCB5843-5G
6-Bromochroman-4-one ≥98.0% (by GC)
6-Bromochroman-4-one
Formula: C₉H₇BrO₂
MW: 227.0547 g/mol
MDL Number: MFCD00546803
CAS Number: 49660-57-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR