tert-Butyl-3-formylpyrrolidine-1-carboxylate ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (+/-)-3-Formylpyrrolidine-1-carboxylic acid tert-Butyl ester, (+/-)-1-Boc-pyrrolidine-3-carboxaldehyde, (+/-)-1-(tert-Butoxycarbonyl)-3-formylpyrrolidine, (+/-)-1-Boc-3-formylpyrrolidine, tert-Butyl 3-formylpyrrolidine-1-carboxylate, (+/-)-tert-Butyl 3-Formylpyrrolidine-1-carboxylate, (+/-)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxaldehyde

B6091-1G B6091-5G
TCB6091-1GEA 92 USD
TCB6091-1G TCB6091-5G
tert-Butyl-3-formylpyrrolidine-1-carboxylate ≥96.0% (by GC)
tert-Butyl-3-formylpyrrolidine-1-carboxylate
Formula: C₁₀H₁₇NO₃
MW: 199.2500 g/mol
MDL Number: MFCD03791257
CAS Number: 59379-02-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR