4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

C2774-25G C2774-5G
TCC2774-25GEA 254 USD
TCC2774-25G TCC2774-5G
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline ≥98.0% (by HPLC, total nitrogen)
4-Chloro-6,7-bis(2-methoxyethoxy)quinazoline
Formula: C₁₄H₁₇ClN₂O₄
MW: 312.7500 g/mol
MDL Number: MFCD09751265
CAS Number: 183322-18-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 99.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR