4-Bromo-7-azaindole ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4-Bromo-7-azaindole, 4-Bromo-1H-pyrrolo[2,3-b]pyridine
B6078-1G
B6078-5G
TCB6078-1GEA
48
USD
TCB6078-1G
TCB6078-5G
4-Bromo-7-azaindole ≥98.0% (by GC, titration analysis)
4-Bromo-7-azaindole
Formula:
C₇H₅BrN₂ |
MDL Number:
MFCD08272233 CAS Number: 348640-06-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 177.0 to 181.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...