(2,4-Dichlorobenzyl)methylamine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
2,4-Dichloro-N-methylbenzylamine, N-Methyl-2,4-dichlorobenzylamine
D5147-1G
D5147-5G
TCD5147-1GEA
53
USD
TCD5147-1G
TCD5147-5G
(2,4-Dichlorobenzyl)methylamine ≥98.0% (by GC, titration analysis)
(2,4-Dichlorobenzyl)methylamine
Formula:
C₈H₉Cl₂N MW: 190.07 g/mol Boiling Pt: 123 °C |
MDL Number:
MFCD00045185 CAS Number: 5013-77-4 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...