tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: [(1R,2R)-2-Aminocyclohexyl]carbamic acid tert-Butyl ester, (1R,2R)-2-Amino-1-(Boc-amino)cyclohexane, (1R,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine, (1R,2R)-N-Boc-1,2-cyclohexanediamine, tert-Butyl [(1R,2R)-2-Aminocyclohexyl]carbamate, (1R,2R)-N1-Boc-1,2-cyclohexanediamine, (1R,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane

B5647-1G B5647-5G
TCB5647-1GEA 286 USD
TCB5647-1G TCB5647-5G
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate ≥98.0% (by GC, titration analysis)
tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate
Formula: C₁₁H₂₂N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08726023
CAS Number: 146504-07-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 114.0 to 118.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR