5-Bromo-2-hydroxy-4-picoline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 5-Bromo-2-hydroxy-4-methylpyridine

B4701-25G B4701-5G
TCB4701-25GEA 352 USD
TCB4701-25G TCB4701-5G
5-Bromo-2-hydroxy-4-picoline ≥98.0% (by GC, titration analysis)
5-Bromo-2-hydroxy-4-picoline
Formula: C₆H₆BrNO
MW: 188.02 g/mol
MDL Number: MFCD03427664
CAS Number: 164513-38-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 199.0 to 203.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR