(R)-β-Phenylalaninol ≥97.0% (by GC)

Supplier: TCI America
Danger

Synonyms: (R)-3-Amino-3-phenylpropan-1-ol, (R)-1-Phenyl-3-propanolamine, (R)-3-Amino-3-phenyl-1-propanol

A2999-1G A2999-200MG
TCA2999-1GEA 140 USD
TCA2999-1G TCA2999-200MG
(R)-β-Phenylalaninol ≥97.0% (by GC)
(R)-β-Phenylalaninol
Formula: C₉H₁₃NO
MW: 151.2056 g/mol
Melting Pt: 77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01311791
CAS Number: 170564-98-4
UN: 3259
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 93.0 %
Specific rotation [a]20/D +21.0 to +25.0 deg (C=1, CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR