5'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

B5237-1G B5237-5G
TCB5237-1GEA 35 USD
TCB5237-1G TCB5237-5G
5'-Bromo-2'-fluoroacetophenone ≥98.0% (by GC)
5'-Bromo-2'-fluoroacetophenone
Formula: C₈H₆BrFO
MDL Number: MFCD11226880
CAS Number: 198477-89-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR