5-Bromo-3-nitropyridine-2-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

B5584-25G B5584-5G
TCB5584-25GEA 239 USD
TCB5584-25G TCB5584-5G
5-Bromo-3-nitropyridine-2-carbonitrile ≥98.0% (by GC)
5-Bromo-3-nitropyridine-2-carbonitrile
Formula: C₆H₂BrN₃O₂
MDL Number: MFCD06657551
CAS Number: 573675-25-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR